Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin (Muscle Cell Marker) Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB9014P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9014P1 500 μL Concentrated
RB9014P0 100 μL Concentrated
RB9014P 1 mL Concentrated
RB9014R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB9014P1 Supplier Epredia Supplier No. RB9014P1
Only null left
Add to Cart
Add to Cart
Request a Quote

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Desmin is an intermediate filament protein of both smooth and striated muscles. Antibody to desmin reacts with striated (skeletal and cardiac) as well as smooth muscle cells. In skeletal and cardiac muscles, the staining is confined to the Z-bands giving a characteristic striated appearance. Anti-desmin antibody is useful in identification of tumors of myogenic origin.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Near C-terminus

Immunogen: A synthetic peptide mapping near the C-terminus of human desmin

Molecular Weight: 53 kDa

Positive Control: Muscle or Sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Specifications

Antigen Desmin (Muscle Cell Marker)
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide mapping near the C-terminus of human desmin
Quantity 500 μL
Regulatory Status IVD
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.