Learn More
Desmin (Muscle Cell Marker) Rabbit Polyclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.
Supplier: Epredia RB9014R1
Description
Desmin is an intermediate filament protein of both smooth and striated muscles. Antibody to desmin reacts with striated (skeletal and cardiac) as well as smooth muscle cells. In skeletal and cardiac muscles, the staining is confined to the Z-bands giving a characteristic striated appearance. Anti-desmin antibody is useful in identification of tumors of myogenic origin.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: Near C-terminus
Immunogen: A synthetic peptide mapping near the C-terminus of human desmin
Molecular Weight: 53 kDa
Positive Control: Muscle or Sarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
Desmin (Muscle Cell Marker) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
1 mL | |
Muscle Biology | |
Human, Mouse | |
IgG |
Immunoprecipitation, Western Blot | |
Polyclonal | |
Ready-to-use for IHC (UVLP) | |
A synthetic peptide mapping near the C-terminus of human desmin | |
IVD | |
Primary | |
Prediluted |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.