Learn More
Epredia™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS1898S1
Description
Host Species: Mouse
Clone: Ber-EP4
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: MCF-7 human breast carcinoma cell line
Molecular Weight: ∽34 and 39 kDa (reducing); 39 and 41 kDa (nonreducing); 31 and 36 kDa (deglycosylated)
Positive Control: Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Epithelial Specific Antigen/Ep-CAM Ab-9 | |
| Monoclonal | |
| Unconjugated | |
| MCF-7 human breast carcinoma cell line | |
| IVD | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| Ber-EP4 | |
| Mouse | |
| 500 μL | |
| Cancer and Tumor Biology | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.