Learn More
Epredia™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-9, Mouse Monoclonal Antibody.
$257.00 - $1136.00
Specifications
| Antigen | Epithelial Specific Antigen/Ep-CAM Ab-9 |
|---|---|
| Clone | Ber-EP4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|
MS1898S0
|
Epredia™
MS1898S0 |
Concentrated | 100 μL |
Each for $257.00
|
|
||||
|
MS1898S1
|
Epredia™
MS1898S1 |
Concentrated | 500 μL |
Each for $610.00
|
|
||||
|
MS1898S
|
Epredia™
MS1898S |
Concentrated | 1 mL |
Each for $1,136.00
|
|
||||
|
MS1898R7
|
Epredia™
MS1898R7 |
Prediluted | 7 mL |
Each for $574.00
|
|
||||
|
MS1898R1
|
Epredia™
MS1898R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
Description
Host Species: Mouse
Clone: Ber-EP4
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: MCF-7 human breast carcinoma cell line
Molecular Weight: ∽34 and 39 kDa (reducing); 39 and 41 kDa (nonreducing); 31 and 36 kDa (deglycosylated)
Positive Control: Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Epithelial Specific Antigen/Ep-CAM Ab-9 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG2a κ |
| Ber-EP4 | |
| Monoclonal | |
| Mouse | |
| Cancer and Tumor Biology | |
| MCF-7 human breast carcinoma cell line | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.