Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Epstein-Barr Virus/LMP Ab-1 Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epstein-Barr Virus/LMP Ab-1, Mouse Monoclonal Antibody.

$755.00

Specifications

Antigen Epstein-Barr Virus/LMP Ab-1
Clone CS1+CS2+CS3+CS4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS1458S1
View Documents
Epredia™
MS1458S1A
Concentrated 500 μL
Each for $755.00
Only null left
Add to Cart
 
MS1458S0
View Documents
Epredia™
MS1458S0A
Concentrated 100 μL N/A N/A N/A
MS1458S
View Documents
Epredia™
MS1458SA
Concentrated 1 mL N/A N/A N/A
MS1458R7
View Documents
Epredia™
MS1458R7A
Prediluted 7 mL N/A N/A N/A
Description

Description

Host Species: Mouse

Clone: CS1 + CS2 + CS3 + CS4

Isotype: IgG1/κ

Species Reactivity: EBV-positive cells

Epitope: Located in the internal part of the membrane-associated viral LMP

Immunogen: EBV-encoded recombinant latent membrane protein

Molecular Weight: 60 kDa (LMP)

Positive Control: EBV-positive lymphoblastoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Epstein-Barr Virus/LMP Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
ASR, IVD
EBV-encoded recombinant latent membrane protein
Primary
CS1+CS2+CS3+CS4
Monoclonal
Mouse
Infectious Disease
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.