Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Estrogen Receptor Ab-14, Mouse Monoclonal Antibody

Catalog No. p-4542564 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS750S0 100 μL Concentrated
MS750S1 500 μL Concentrated
MS750S 1 mL Concentrated
MS750R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS750S0 Supplier Epredia™ Supplier No. MS750S0A
Only null left
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Estrogen Receptor Ab-14, Mouse Monoclonal Antibody.

The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Mouse

Clone: 1D5 + 6F11

Isotype: IgG1 + IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant human estrogen receptor protein

Molecular Weight: 67 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen Estrogen Receptor Ab-14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1D5 + 6F11
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human estrogen receptor protein
Quantity 100 μL
Regulatory Status ASR, IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.