Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Follicle Stimulating Hormone (FSH) Ab-3, Mouse Monoclonal Antibody, Epredia™
Epredia™ Follicle Stimulating Hormone (FSH) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.
Specifications
Antigen | Follicle Stimulating Hormone (FSH) Ab-3 |
---|---|
Clone | FSH03 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1449P1ABX
|
Epredia
MS1449P1ABX |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS1449PABX
|
Epredia
MS1449PABX |
Concentrated | 200 μL | N/A | N/A | N/A | |||
MS1449R1
|
Epredia
MS1449R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
MS1449R7
|
Epredia
MS1449R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
FSH is a pituitary hormone involved in the maturation of ovarian follicles and estrogen secretion in females. In the pituitary gland, FSH is produced by gonadotrophs. In males, FSH stimulates the secretion of testosterone.Host Species: Mouse
Clone: FSH03
Isotype: IgG1/κ
Species Reactivity: Human and Cow. Others not known.
Immunogen: Purified human FSH-β
Positive Control: Anterior pituitary
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Follicle Stimulating Hormone (FSH) Ab-3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Bovine, Human | |
IgG2a κ |
FSH03 | |
Monoclonal | |
Mouse | |
Steroids Hormones and Growth Factors | |
Purified human FSH-Beta | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title