Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Ku (p70/p80) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Recommended for multiple molecular biology techniques, Epredia™ Ku (p70/p80) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

Supplier:  Epredia™ MS286R1

 View more versions of this product

Catalog No. MS286R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

The Ku autoantigen is a heterodimer of 70kDa (p70) and ∽80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.

Host Species: Mouse

Clone: 162

Isotype: IgG2a

Species Reactivity: Human, Monkey, Xenopus. Mouse and Rat (Immunoprecipitation only). Does not react with cow and rabbit.

Epitope: A conformational epitope of p70/p80 dimer, which is destroyed during Western Blotting

Immunogen: Human B-cell nuclei from plasmacytoid 2p68 cells

Molecular Weight: 70kDa and 80kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Affinity Purification
  • Flow Cytometry
  • Gel Supershift

General References:

1. Wang J, et. al. Molecular Biology Reports, 1993, 18(1):15-28.

2. Reeves WH, et. al. Journal of Experimental Medicine, 1985, 161(1):18-39.

Specifications

Specifications

Ku (p70/p80) Ab-3
Monoclonal
Unconjugated
Mouse
1 mL
DNA Repair
Human, Non-human Primate, Xenopus species
IgG2a
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
162
Ready-to-use for IHC (UVLP)
Human B cell nuclei from plasmacytoid 2p68 cells
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.