Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Laminin Ab-1 Rabbit Polyclonal Antibody with BSA and azide

Recommended for Immunohistochemistry (Formalin/paraffin). Epredia™ Laminin Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Specifications

Antigen Laminin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RB082A0
View Documents
Epredia™
RB082A0
Concentrated 100 μL N/A N/A N/A
RB082A
View Documents
Epredia™
RB082A
Concentrated 1 mL N/A N/A N/A
RB082R1
View Documents
Epredia™
RB082R1
Prediluted 1 mL N/A N/A N/A
RB082RQ
View Documents
Epredia™
RB082RQ
Ready-to-Use 12 mL N/A N/A N/A
Description

Description

Extracellular matrix (ECM) surrounds and separates most tissue compartments, determines tissue architecture and dynamically interacts with its surrounding cells. The basement membrane is a part of the ECM and is composed of collagens, proteoglycans, elastin and several glycoproteins of which laminin is the most abundant. Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains Dystroglycan is a major laminin receptor in muscle and epithelia, and nidogen is a high-affinity laminin-binding protein important for basement-membrane assembly.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Laminin isolated from a rat yolk sac tumor cell line

Positive Control: Skin, Lung, Kidney or Tonsil

Cellular Localization: Basement membrane Ab-1 labels basement membrane surrounding normal vessels, glands, muscles, nerves, fat and deciduas cells separating these from abutting connective tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Laminin Ab-1
Polyclonal
Rabbit
Extracellular Matrix
Primary
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Laminin isolated from a rat yolk sac tumor cell line
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.