Learn More
MUC-1 Rabbit Polyclonal Antibody, Epredia™
MUC-1, Rabbit Polyclonal Ab is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Supplier: Epredia RB9222R1
Description
MUC-1 is a heavily O-glycosylated transmembrane protein expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and overexpressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC-1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Mouse
Epitope: aa 239-255
Immunogen: A synthetic peptide from cytoplasmic tail of human MUC-1 protein
Molecular Weight: 265-400kDa
Positive Control: MCF-7 cells; Breast CA
Cellular Localization: Cytoplasmic and membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| MUC-1 | |
| Polyclonal | |
| Rabbit | |
| 1 mL | |
| Cancer and Tumor Biology | |
| Human | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| A synthetic peptide from cytoplasmic tail of human MUC-1 protein | |
| IVD | |
| Primary | |
| Prediluted |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.