Learn More
MUC-1 Rabbit Polyclonal Antibody, Epredia™
MUC-1, Rabbit Polyclonal Ab is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Specifications
| Antigen | MUC-1 |
|---|---|
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|
RB9222P1
|
Epredia
RB9222P1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
|
RB9222P
|
Epredia
RB9222P |
Concentrated | 1 mL | N/A | N/A | N/A | |||
|
RB9222R1
|
Epredia
RB9222R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
|
RB9222R7
|
Epredia
RB9222R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
MUC-1 is a heavily O-glycosylated transmembrane protein expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and overexpressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC-1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Mouse
Epitope: aa 239-255
Immunogen: A synthetic peptide from cytoplasmic tail of human MUC-1 protein
Molecular Weight: 265-400kDa
Positive Control: MCF-7 cells; Breast CA
Cellular Localization: Cytoplasmic and membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| MUC-1 | |
| Polyclonal | |
| Rabbit | |
| Cancer and Tumor Biology | |
| A synthetic peptide from cytoplasmic tail of human MUC-1 protein | |
| Primary |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.