Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC-1 Rabbit Polyclonal Antibody, Epredia™

MUC-1, Rabbit Polyclonal Ab is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Supplier:  Epredia RB9222R7

 View more versions of this product

Catalog No. RB9222R7


Description

Description

MUC-1 is a heavily O-glycosylated transmembrane protein expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and overexpressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC-1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse

Epitope: aa 239-255

Immunogen: A synthetic peptide from cytoplasmic tail of human MUC-1 protein

Molecular Weight: 265-400kDa

Positive Control: MCF-7 cells; Breast CA

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

MUC-1
Polyclonal
Rabbit
7 mL
Cancer and Tumor Biology
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
A synthetic peptide from cytoplasmic tail of human MUC-1 protein
IVD
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.