Learn More
Epredia™ Lab Vision™ Myosin, Smooth Muscle Heavy Chain, Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Provides accurate, reproducible results with the Epredia™ Myosin, Smooth Muscle Heavy Chain Ab-1, Mouse Monoclonal Antibody.
Specifications
Antigen | Myosin, Smooth Muscle Heavy Chain Ab-1 |
---|---|
Clone | SMMS-1 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1177P0
|
Epredia™
MS1177P0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS1177P1ABX
|
Epredia™
MS1177P1ABX |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS1177PABX
|
Epredia™
MS1177PABX |
Concentrated | 200 μL | N/A | N/A | N/A | |||
MS1177P1
|
Epredia™
MS1177P1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
MS1177R1
|
Epredia™
MS1177R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
MS1177RQ
|
Epredia™
MS1177RQ |
Ready-to-Use | 12 mL | N/A | N/A | N/A | |||
Description
SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.Host Species: Mouse
Clone: SMMS-1
Isotype: IgG1/k
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Crude human uterus extract
Molecular Weight: 205kDa (MHC-1) and 200kDa (MHC-2)
Positive Control: Uterus
Cellular Localization: Cytoplasmic
Recommended for:
Immunohistochemistry (Formalin/paraffin)
Specifications
Myosin, Smooth Muscle Heavy Chain Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Crude human uterus extract |
SMMS-1 | |
Monoclonal | |
Mouse | |
Muscle Biology | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.