Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ NFκB/p65 (Rel A) Ab-1 Rabbit Polyclonal Antibody with BSA and azide

Epredia™ NFκB/p65 (Rel A) Ab-1, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Supplier:  Epredia™ RB1638R1

 View more versions of this product

Catalog No. RB1638R1


Description

Description

NFκ B is a heterodimer consisting of a 50kDa DNA binding subunit and a 65kDa transactivating subunit (RelA). The p50/p65 heterodimer is present in the cytoplasm in inactive form bound to its inhibitor, IdeltaB. Upon stimulation of cells with stimuli like LPS, Interleukin-1, IkB gets phosphorylated and degraded by proteosomes releasing the activated NFκB which goes to the nucleus inducing gene expression. NFκB was originally identified as transcriptional activator for kappa light chain gene in B cells.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Hamster, Rabbit, Cow, Pig, Dog, Drosophila, Xenopus. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of human NFκB protein

Molecular Weight: 65kDa

Positive Control: Jurkat cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

NFkappaB/p65 (Rel A) Ab-1
Polyclonal
Ready-to-use for IHC (UVLP)
A synthetic peptide from the C-terminus of human NFkappaB protein
RUO
Primary
Prediluted
Western Blot
Unconjugated
Rabbit
1 mL
Transcription Regulation
Bovine, Canine, Drosophila, Hamster, Human, Mouse, Pig, Rabbit, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.