Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X)

Catalog No. ta125pt Shop All Epredia Products
Click to view available options
Quantity:
125 mL
1000 mL

Employ a wash buffer for immunolabeling techniques with the Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X).

  • For in vitro diagnostic use
  • Added to promote more effective washings, resulting in decreased nonspecific background staining
  • Aids reagent spreading on slides

Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week.

Specifications

Chemical Name or Material PBS and Tween
Concentration 20X
For Use With (Application) Anatomical Pathology
pH 7.6
Quantity 125 mL

For in vitro diagnostic use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.