Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Prolactin Mouse Monoclonal Antibody with BSA and azide

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Prolactin, Mouse Monoclonal Antibody.

Specifications

Antigen Prolactin
Clone SPM108
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS9083P0
View Documents
Epredia™
MS9083P0
Concentrated 100 μL N/A N/A N/A
Description

Description

Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. Prolactin producing cells make up approximately 20% of the pituitary.

Host Species: Mouse

Clone: SPM108

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Immunogen: Purified human prolactin

Molecular Weight: 23.5kDa

Positive Control: Pituitary gland

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Prolactin
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human
IgG1 κ
SPM108
Monoclonal
Mouse
Steroids Hormones and Growth Factors
Purified human prolactin
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.