Learn More
Epredia™ Lab Vision™ S100 Rabbit Polyclonal Antibody with BSA and azide
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ S100, Rabbit Polyclonal Antibody.
Specifications
Antigen | S100 |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
RB9018P
|
Epredia™
RB9018P |
Concentrated | 1 mL | N/A | N/A | N/A | |||
RB9018R1
|
Epredia™
RB9018R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
RB9018RQ
|
Epredia™
RB9018RQ |
Ready-to-Use | 12 mL | N/A | N/A | N/A | |||
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. Antibody to S100 stains Schwannomas, ependymomas, astrogliomas, almost all benign and malignant melanomas and their metastases. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant protein encoding human S100 protein
Molecular Weight: 21-24kDa
Positive Control: Melanoma or Schwannoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
S100 | |
Polyclonal | |
Rabbit | |
Cancer and Tumor Biology | |
Primary |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Recombinant protein encoding human S100 protein |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.