Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ S100A4 Ab-8 Rabbit Polyclonal Antibody with BSA and azide

Epredia™ S100 A4 Ab-8, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Supplier:  Epredia™ RB1804R1

 View more versions of this product

Catalog No. RB1804R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

The S100A4 protein belongs to the S100 subfamily, now designated S100A1/S100A13. This antibody does not cross react with other S100 proteins. Anti-S100A4 labels fibroblasts, neuronal structures, T-lymphocytes, plasma cells and Kupffer cells in normal tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant human S100 A4 monomer expressed in E. coli

Molecular Weight: 10-12kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic, membrane and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

S100 A4 Ab-8
Polyclonal
Unconjugated
Purified recombinant human S100 A4 monomer expressed in E. coli
RUO
Primary
Prediluted
Immunohistochemistry (Paraffin)
Polyclonal
Rabbit
1 mL
Cancer and Tumor Biology
Bovine, Canine, Equine, Feline, Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.