Learn More
Epredia™ Lab Vision™ S100A4 Ab-8 Rabbit Polyclonal Antibody with BSA and azide
Epredia™ S100 A4 Ab-8, Rabbit Polyclonal Antibody provides accurate, reproducible results.
Specifications
| Antigen | S100 A4 Ab-8 |
|---|---|
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|
RB1804R1
|
Epredia™
RB1804R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
Description
The S100A4 protein belongs to the S100 subfamily, now designated S100A1/S100A13. This antibody does not cross react with other S100 proteins. Anti-S100A4 labels fibroblasts, neuronal structures, T-lymphocytes, plasma cells and Kupffer cells in normal tissues.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant human S100 A4 monomer expressed in E. coli
Molecular Weight: 10-12kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic, membrane and nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| S100 A4 Ab-8 | |
| Polyclonal | |
| Rabbit | |
| Cancer and Tumor Biology | |
| Purified recombinant human S100 A4 monomer expressed in E. coli | |
| Primary |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Bovine, Canine, Equine, Feline, Human, Mouse | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.