Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Tartrate Resistant Acid Phosphatase (TRAP) Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1768S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1768S1 500 μL Concentrated
MS1768S0 100 μL Concentrated
MS1768S 1 mL Concentrated
MS1768R1 1 mL Prediluted
MS1768R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1768S1 Supplier Epredia™ Supplier No. MS1768S1
Add to Cart
Add to Cart

Tartrate Resistant Acid Phosphatase (TRAP) Ab-1, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.

Host Species: Mouse

Clone: 26E5

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein corresponding to N-terminal region of human TRAP

Positive Control: Lung (macrophages)

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Tartrate Resistant Acid Phosphatase (TRAP) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 26E5
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminal region of human TRAP
Purification Method Lung (macrophages)
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.