Learn More
Epredia™ Lab Vision™ Tartrate Resistant Acid Phosphatase (TRAP) Ab-1, Mouse Monoclonal Antibody
Tartrate Resistant Acid Phosphatase (TRAP) Ab-1, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
$1867.00
Specifications
| Antigen | Tartrate Resistant Acid Phosphatase (TRAP) Ab-1 |
|---|---|
| Clone | 26E5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Form | Price | Quantity & Availability | ||||
|
MS1768S1
|
Epredia™
MS1768S1 |
500 μL | Concentrated |
Each for $1,867.00
|
|
||||
|
MS1768S0
|
Epredia™
MS1768S0 |
100 μL | Concentrated | N/A | N/A | N/A | |||
|
MS1768S
|
Epredia™
MS1768S |
1 mL | Concentrated | N/A | N/A | N/A | |||
|
MS1768R1
|
Epredia™
MS1768R1 |
1 mL | Prediluted | N/A | N/A | N/A | |||
|
MS1768R7
|
Epredia™
MS1768R7 |
7 mL | Prediluted | N/A | N/A | N/A | |||
Description
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.Host Species: Mouse
Clone: 26E5
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: N-terminal
Immunogen: Recombinant protein corresponding to N-terminal region of human TRAP
Positive Control: Lung (macrophages)
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Tartrate Resistant Acid Phosphatase (TRAP) Ab-1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG2b |
| 26E5 | |
| Monoclonal | |
| Mouse | |
| Cancer and Tumor Biology | |
| Recombinant protein corresponding to N-terminal region of human TRAP | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.