Learn More
Labconco™ Atmospure Cable
Shop All Labconco Corporation Products
Click to view available options
Product Type:
Atmospure 6-Pin Cable
Atmospure 9-Pin Cable
Description
AtmosPure 6-Pin Communication Cable is required for use when AtmosPure Re-Gen Gas Purifiers and glove box configured with Auto Pressure Controller are sharing one vacuum pump.
Specifications
Specifications
| For Use With (Equipment) | AtmosPure |
| Weight (English) | 2 lb. |
| Weight (Metric) | 0.9 Kg |
| Product Type | Atmospure 6-Pin Cable |
| For Use With (Application) | For use when Atmospure and glove box with auto pressure controller are sharing one vacuum pump |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov