Learn More
Labconco™ Freeze-Drying Ampule Accessories
SDP
Catalog No. p-21252 Shop All Labconco Corporation Products
Change view
Click to view available options
Type:
Ampule Adapter
Sealing Torch
Stopper
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
10-269-81B Stopper
10-269-82 Sealing Torch
10-269-80 Ampule Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-269-81B Supplier Labconco™ Supplier No. 7576210
Only null left
Add to Cart
Add to Cart
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
  • Large rubber split stoppers fit 5, 10, 20, 30, 50, 100 and 125 ml Labconco Serum Bottles (bottles with 20 mm corkage).
    • Package of 100.
  • Oxygen/Natural Gas Sealing Torch is specifically designed for flame sealing freeze dry ampules.
    • Seals all types of heat-resistant glass.
    • Connects to natural gas, butane or propane and oxygen with 1/4 in. ID hose connectors.
  • Ampule valve adapter provides connection of ampules to drying chambers and manifolds by connecting ampules to valve ports or stainless steel stems.
    • Ten per package.
Specifications
Type Stopper
Material Rubber
Quantity 100 Pk.
For Use With (Equipment) Freeze drying system
Diameter (Metric) 2 cm