Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ Protector™ Double Stainless Steel Combination Glove Boxes
Shop All Labconco Corporation Products

Click to view available options
Certifications/Compliance:
CE
UL, ETL, CAN/CSA
Plug Type:
China/Australia Plug
North America, 100/115 V
North America, 208/230 V
Schuko Plug
UK Plug
Voltage:
100/115 V
208/230 V
Description
Includes
Auto-Pressure Controller
- Inner and outer pivoting transfer chamber doors, counterbalanced and equipped with quick-latches
- Two rear-mounted inlet and outlet 0.75 in. OD compression fittings for connection to optional AtmosPure™ Re-Gen Gas Purifier or other accessory drying train
- Interior right-hand side wall-mounted electrical receptacle (duplex on 100/115V models) with 10A maximum
- Interior left-hand side wall-mounted pass-through with plug
- Exceed Class 1 atmosphere containment conditions for oxygen permeation <1.67ppm/min per ISO 10648-2 test method
- Centrally-mounted electrical receptacle
- Type 304 stainless steel liner, Type 304 stainless steel inner and outer transfer chamber doors and Type 304 stainless steel front panel
- Four fluorescent lights with two viewing windows and Four neoprene gloves and glove ports
- Illumination exceeds 80 footcandles
- Work with both atmosphere-sensitive and/or hazardous materials.
Specifications
Specifications
| Product Type | Glove Box |
| Filter Type | ULPA |
| Lighting Type | Fluorescent (2 x 25 W) |
| Material | Stainless Steel |
| Amperage | 10 A |
| Certifications/Compliance | CE |
| Depth (English) | 34.4 in. |
| Depth (Metric) | 87.4 cm |
| Display | LCD |
| For Use With (Application) | Laboratories that have multiple types of glove box applications or anticipate that their applications may change in the future (atmosphere sensitive materials and hazardous materials) |
| Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction