Learn More
Labconco™ Protector Glove Box Accessory Kits
SDP
Kits provided added functionality options for Protector glove boxes
$1238.00 - $1551.00
Specifications
For Use With (Equipment) Protector Glove Boxes
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
16-200-386
SDP
View Documents
Labconco™
5442700
Add-A-Valve Kit for Main Chamber
Each for $1,551.00
Only null left
Add to Cart
16-200-387
SDP
View Documents
Labconco™
5441403
Pressure Relief Kit
Each for $1,238.00
Only null left
Add to Cart
Description
Description
Add-A-Valve Kits include two manual valves, tubes, nuts and instructions for adding valves to the glove box for evacuating with vacuum, filling with inert gas, connecting to a drying train, or connecting to filtration components. Manual valves mount in predrilled holes behind the label located below the control panel. For use with Protector Combination and Protector Filtered Glove Boxes.
Specifications
Specifications
Protector Glove Boxes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.