Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™ Standard Storage Cabinet
SDP

Catalog No. 16080321
Click to view available options
Width (English):
12 in.
18 in.
24 in.
30 in.
36 in.
48 in.
No. of Doors:
1
2
Door Type:
Left Hinge, Manual Close
Manual Close
Right Hinge, Manual Close
Weight (English):
60 lb.
65 lb.
85 lb.
90 lb.
120 lb.
130 lb.
155 lb.
180 lb.
260 lb.

Includes

Four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in.

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

  • Support loads up to 800 lb. (363kg)
  • Durable epoxy-coated steel construction
  • Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
  • Manual-closing, non-locking door(s)

Order Info

Work surface and interior shelf kits are sold separately

Specifications

Product Type Storage Cabinet
Color Light Gray
Door Type Right Hinge, Manual Close
No. of Doors 1
Weight (English) 85 lb.
Weight (Metric) 38.55 kg
Height (English) Base 31.5 to 32.75 in.
Includes Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
Load Bearing Capacity (English) 800 lb.
Load Bearing Capacity (Metric) 362.8 kg
Height (Metric) Base 80 to 83.18 cm
Width (English) 18 in.
Width (Metric) 45.72 cm
For Use With (Application) Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
Certifications/Compliance ADA
Depth (English) 22 in.
Depth (Metric) 55.88 cm
Material Epoxy-coated Steel
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.