Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Vertical Clean Bench
SDP

Catalog No. 16108267 Shop All Labconco Corporation Products
Click to view available options
Width (English):
24 in.
36 in.
48 in.
Voltage:
115 V
230 V
Plug Type:
No Plug
North America Plug
Includes:
Airflow Monitor
Bench Only
UV, Panel, Timer
UV, Panel, Timer, Airflow Monitor
Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Catalog No. 16-108-267

Supplier: Labconco™ 3970424

Only null left
Add to Cart
Add to Cart
Request a Quote
Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Requires

Work surface and base stand (sold separately).

Protect work from particulate contamination by choosing a Purifier™ Vertical Clean Bench.

  • Nominal downflow velocity of 45-65 fpm and aerodynamic air foil
  • Supply HEPA filter, 99.99% efficient on particles 0.3 Micron
  • ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
  • Variable speed built-in blower with motorized impeller and solid state control rated for 10 A
  • Intrinsically-sound, negative pressure design
  • Minihelic pressure gauge and disposable prefilter
  • UV-resistant, glacier white and gray, powder-coated aluminum and steel construction
  • UV-opaque and resistant 6.3mm (0.25 in.) thick tempered safety glass sash and sides
  • Light and blower switches
  • Air diffuser and 23.8cm (9.4 in.) high sash opening
  • Ergonomic 10° angled and hinged sash that pivots upward, locks to a loading height of 50.8cm (20 in.) and has wiping seals to promote ISO Class 5 conditions
  • Two utility ports with iris openings and plugs
  • ETL Listing (115 volt, 60 Hz models)

Order Info

Vertical Clean Benches can be ordered with bifold doors; specify when ordering. Guardian™ Airflow Monitor is offered as factory-installed option on all benches; specify when ordering.

Warranty and Services

One year parts and service

Specifications

Product Type Clean Bench
Coating Powder
Filter Type HEPA
Lighting Type Fluorescent
Material Powder-coated Aluminum and Steel
Amperage 3 A
Certifications/Compliance CAN/CSA C22.2, UL, CE
Color Glacier White, Gray
Depth (English) 28.6 in.
Depth (English) Interior 25.3 in.
Depth (Metric) 72.64 cm
Depth (Metric) Interior 64.3 cm
For Use With (Application) Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation
Height (English) 36.8 in.
Height (English) Interior 22.7 in.
Height (Metric) 93.47 cm
Height (Metric) Interior 57.7 cm
Hertz 50/60 Hz
Includes UV, Panel, Timer, Airflow Monitor
IP Rating None
Plug Type No Plug
Ports Utility
Voltage 230 V
Weight (English) 195 lb.
Weight (Metric) 88 kg
Width (English) 48 in.
Width (English) Interior 46.8 in.
Width (Metric) 121.9 cm
Width (Metric) Interior 118.9 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.