Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ VACUUBRAND™ Hybrid™ Vacuum Pumps
GSA_VASDP

Catalog No. p-4393940 Shop All Labconco Corporation Products
Change view
Click to view available options
Frequency:
50/60 Hz
60 Hz
Voltage:
115 V
230 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frequency Voltage
10-269-60 60 Hz 115 V
10-030-182 50/60 Hz 230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-269-60 Supplier Labconco™ Supplier No. 7584000
Only null left
Add to Cart
Add to Cart
Request a Quote

Integrates high performance rotary vane pump with a chemical-resistant diaphragm pump to create a low maintenance, longer-lasting pump

Designed for use with acids and other chemicals including TFA, TFA by-products, acetonitrile, HBr and HNO3, present in samples such as HPLC-prepared and peptide purified materials.

  • Combination pump system consists of a two-stage rotary vane pump and two-stage, chemically-resistant diaphragm pump
  • Pressure control valve compensates for the different volumes displaced by the two pumps
  • Ultimate vacuum (partial pressure) 3 x 10(-3)mBar (2.25 micron)
  • Ready to use and fully charged with vacuum pump oil (approximately 1L)
  • 0.75 in. OD inlet adapter
  • Refill pump oil sold separately, 10-030-181

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Max. Vacuum <3 x 10-3 mBar
Amperage 5.1 A
Includes 1L Bottle of vacuum pump Oil, power switch, power cord and plug, 0.75 in. O.D. inlet adapter
Electrical Requirements 115 V 60 Hz
Dimensions (L x W x H) 9 x 18.9 x 15 in.
Noise Level 55 dB
Weight (English) 66 lb.
Weight (Metric) 30 kg
Flow Rate 115 L/min.
Frequency 60 Hz
Overall Height (English) 15 in.
Overall Height (Metric) 38.1 cm
Overall Length (English) 9 in.
Overall Length (Metric) 22.9 cm
Overall Width (English) 18.9 in.
Pump Type Vacuum Pump
Maximum Pressure 1 atm
Voltage 115 V
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.