Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio Label IT™ Tracker™ Intracellular Nucleic Acid Localization Kit, Biotin
Shop All Mirus Bio LLC Products
Click to view available options
:
Localization Kit, Biotin
Description
- Versatile Labeling: Efficiently label and visualize the DNA of your choice
- One-step Chemical Method: Easily and consistently control the labeling reactions
- High Efficiency Labeling: Optimal visualization of DNA in cells
- Superior Tracking: Monitor both subcellular localization and reporter transgene expression following transfection of labeled (plasmid) DNA
Specifications
Specifications
Format | Kit |
Label or Dye | Biotin |
Final Product Type | DNA (Labeled), RNA (Labeled) |
Labeling Target | DNA/RNA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction