Learn More
United Scientific™ Laboratory Jack
SDPSCI_ED
Easily adjust with the simple turn of a knob. The Laboratory Jack is constructed of a durable combination of chemically resistant anodized aluminum and stainless steel.
Supplier:  United Scientific™ LBJK12
 View more versions of this product
Catalog No. S00643
Only null left
Add to Cart
Alert:
Prop 65 Warning
Description
Description
  • Constructed of chemically resistant anodized aluminum and stainless steel.
  • The scissor jack design enables height adjustment with a simple turn of the knob.
Specifications
Specifications
Anodized Aluminum, Stainless Steel
Closed: 4 in., Maximum: 19.75 in.
12 in.
12 in.
99 lb.
Laboratory Jack
10.16 cm (Closed), 50.16 cm (Maximum)
305 x 305 mm (12 x 12 in.)
305 mm
305 mm
45 kg
Product Suggestions
Product Suggestions
Videos
FDS
Documents
Documents
Product Certifications
Promotions
Promotions
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'
missing translation for 'thankYou' missing translation for 'feedbackSubmitted'