Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Lac-Phe

Catalog No. p-200113247
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Metabolite generated by cytosol nonspecific dipeptidase (CNDP2) during intense exercise

Lac-Phe is a metabolite generated from lactate and phenylalanine by cytosolic non-specific dipeptidase (CNDP2) during intense exercise. Lac-Phe reduces food intake in diet-induced obese mice without affecting movement or energy expenditure. When administrated chronically, Lac-Phe decreases adiposity and body weight and improves glucose homeostasis.

Specifications

Chemical Name or Material N-[(2S)-2-Hydroxy-1-oxopropyl]-L-phenylalanine
CAS 183241-73-8
Quantity 50 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C12H15NO4
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.