Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lactamase, beta 2 Antibody (OTI2F9), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N271901P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271901P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271901P594 Supplier Novus Biologicals Supplier No. NBP271901PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

lactamase, beta 2 Monoclonal specifically detects lactamase, beta 2 in Human, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen lactamase, beta 2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2F9
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias beta-lactamase-like protein 2, CGI-83, EC 3.-, lactamase, beta 2
Gene Symbols LACTB2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 78-288 of human lactamase, beta 2 (NP_057111) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51110.0
Target Species Human, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.