Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase A/LDHA Antibody (1A4), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP74023549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP74023549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP74023549 Supplier Novus Biologicals Supplier No. NBP174023JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lactate Dehydrogenase A/LDHA Monoclonal specifically detects Lactate Dehydrogenase A/LDHA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen LactateDehydrogenaseA/LDHA
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1A4
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59
Gene Symbols LDHA
Host Species Mouse
Immunogen Recombinant human LDHA (1-332aa) purified from E.coli (NP_005557)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Mitochondrial Mediated Pathway, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3939
Test Specificity Anti-human LDHA mAb, clone 1A4, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human LDHA protein.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.