Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase A/LDHA Antibody (2066A), DyLight 488, R&D Systems™
SDP

Catalog No. MAB9216G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9216G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB9216G Supplier Novus Biologicals Supplier No. MAB9216G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Lactate Dehydrogenase A/LDHA Monoclonal specifically detects Lactate Dehydrogenase A/LDHA in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen Lactate Dehydrogenase A/LDHA
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2066A
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59
Gene Symbols LDHA
Host Species Rabbit
Immunogen rhLDH-A aa2-92
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Mitochondrial Mediated Pathway, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3939
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.