Learn More
Lactate Dehydrogenase A/LDHA Antibody, DyLight 405, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Lactate Dehydrogenase A/LDHA |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Accession No. | P00338 |
| Gene Alias | Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59 |
| Gene Symbols | LDHA |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.