Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase A/LDHA Antibody (OTI2D11), PerCP, Novus Biologicals™
SDP

Catalog No. NB289072 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289072 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289072 Supplier Novus Biologicals Supplier No. NBP271121PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lactate Dehydrogenase A/LDHA Monoclonal antibody specifically detects Lactate Dehydrogenase A/LDHA in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Lactate Dehydrogenase A/LDHA
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2D11
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59
Host Species Mouse
Immunogen Full length human recombinant protein of human LDHA (NP_005557) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Mitochondrial Mediated Pathway, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3939
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.