Learn More
Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Gene ID (Entrez) | 3945 |
| Gene Symbol | LDHB |
| Quantity | 96 Tests |
| Synonym | EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5 |
| Research Category | HIF Target Genes, Hypoxia, Tumor Biomarkers |
| Species | Non-species specific |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
