Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase B Antibody (2057D), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB295508 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295508 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295508 Supplier Novus Biologicals Supplier No. FAB9205T

Rabbit Monoclonal Antibody

Lactate Dehydrogenase B Monoclonal antibody specifically detects Lactate Dehydrogenase B in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, KnockDown

Specifications

Antigen Lactate Dehydrogenase B
Applications Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 2057D
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5
Host Species Rabbit
Immunogen E. coli-derived recombinant human LDHB, Met1-Val93, Accession # P07195
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline HIF Target Genes, Hypoxia, Mitochondrial Mediated Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3945
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.