Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase B Rabbit anti-Human, Rat, Clone: 6B5I2, Novus Biologicals™
SDP

Catalog No. NB168230 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168230 20 μg
NB168229 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168230 Supplier Novus Biologicals Supplier No. NBP31656920UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Lactate Dehydrogenase B Monoclonal antibody specifically detects Lactate Dehydrogenase B in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Lactate Dehydrogenase B
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6B5I2
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 to 4 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Lactate Dehydrogenase B (P07195).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline HIF Target Genes, Hypoxia, Mitochondrial Mediated Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3945
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.