Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase C Antibody (9F2), Novus Biologicals™
SDP

Catalog No. NB298631 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298631 100 μL
NB298630 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298631 Supplier Novus Biologicals Supplier No. NBP326173100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Lactate Dehydrogenase C Monoclonal antibody specifically detects Lactate Dehydrogenase C in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen Lactate Dehydrogenase C
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 9F2
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Cancer/testis antigen 32, CT32EC 1.1.1.27, EC 1.1.1, lactate dehydrogenase C, lactate dehydrogenase C4, LDH testis subunit, LDH3MGC111073, LDH-C, LDHX, LDH-X, L-lactate dehydrogenase C chain
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Lactate Dehydrogenase C [UniProt P07864]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3948
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.