Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactate Dehydrogenase C Antibody (9F2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Lactate Dehydrogenase C |
---|---|
Clone | 9F2 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
Lactate Dehydrogenase C Monoclonal antibody specifically detects Lactate Dehydrogenase C in Human samples. It is validated for Flow Cytometry, ELISASpecifications
Lactate Dehydrogenase C | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cancer/testis antigen 32, CT32EC 1.1.1.27, EC 1.1.1, lactate dehydrogenase C, lactate dehydrogenase C4, LDH testis subunit, LDH3MGC111073, LDH-C, LDHX, LDH-X, L-lactate dehydrogenase C chain | |
A synthesized peptide derived from Human Lactate Dehydrogenase C [UniProt P07864] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
9F2 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Cellular Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3948 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title