Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase C Antibody, Novus Biologicals™
SDP

Catalog No. NB10068160 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068160 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068160 Supplier Novus Biologicals Supplier No. NB10068160
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Lactate Dehydrogenase C Polyclonal specifically detects Lactate Dehydrogenase C in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen Lactate Dehydrogenase C
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3-1ug/ml, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1ug/ml, Peptide ELISA detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_002292.1
Gene Alias Cancer/testis antigen 32, CT32EC 1.1.1.27, EC 1.1.1, lactate dehydrogenase C, lactate dehydrogenase C4, LDH testis subunit, LDH3MGC111073, LDH-C, LDHX, LDH-X, L-lactate dehydrogenase C chain
Gene Symbols LDHC
Host Species Goat
Immunogen Peptide with sequence C-DSDKEHWKNVHKQ corresponding to internal region according to NP_002292.1, NP_059144.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3948
Test Specificity Both variants represent identical product (NP_002292.1 and NP_059144.1).
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.