Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactoferrin Antibody (006), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB067974 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067974 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067974 Supplier Novus Biologicals Supplier No. NBP289794JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen Lactoferrin
Applications ELISA
Classification Monoclonal
Clone 6
Conjugate Janelia Fluor 549
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Lactoferrin (P02788; Met 1-Lys 710).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules
Primary or Secondary Primary
Gene ID (Entrez) 4057
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.