Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactoferrin Antibody (1A1), Novus Biologicals™
SDP

Catalog No. NB001208 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001208 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001208 Supplier Novus Biologicals Supplier No. NBP1402690.2MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Lactoferrin
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A1
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 10 μg/mL
Formulation PBS (pH 7.4)
Gene Accession No. P02788
Gene Alias EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin
Gene Symbols LTF
Host Species Mouse
Immunogen A purified lactoferrin from human milk.
Purification Method Affinity Purified
Quantity 0.2 mg
Research Discipline Amino Acids Drugs and other small molecules
Gene ID (Entrez) 4057
Test Specificity Recognizes human lactoferrin, a 78kD globular protein found in body secretions such as milk and saliva and is a member of the transferrin family proteins. Transferrins control the level of free iron in the body, by binding and transporting iron. Lactoferrin does not appear to have a transporting function, instead it has antimicrobial activity, by keeping the levels of iron in body fluids low and thus preventing microbes from acquiring iron. Removing free iron from body fluids and inflamed areas also serve to protect against the damaging effects of free radicals. Lactoferrin is used as a therapeutic and prophylactic treatment of various illnesses, such as cancers and intestinal illnesses. May be used to detect lactoferrin in ELISA assays.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.