Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactoferrin Antibody (LF5-1D2), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP267260A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP267260A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP267260A32 Supplier Novus Biologicals Supplier No. NBP267260AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Flow Cytometry, ELISA, Flow (Intracellular).

Specifications

Antigen Lactoferrin
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone LF5-1D2
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, ELISA, Flow (Intracellular)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin
Gene Symbols LTF
Host Species Mouse
Immunogen Human lactoferrin
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Amino Acids Drugs and other small molecules
Primary or Secondary Primary
Gene ID (Entrez) 4057.0
Test Specificity The antibody MEM-61 recognizes an epitope on second extracellular domain (EC2) of CD9 antigen, a 24 kDa transmembrane protein expressed on platelets, monocytes, pre-B lymphocytes, granulocytes and activated T lymphocytes.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.