Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaemmLi SDS Sample Buffer, 6X, Reducing, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
Description
General Description
- This compound is composed of several compounds, including 375 mM Tris-HCl (pH 6.8), 9% SDS, 50% glycerol, 9% B-mercaptoethanol, 0.03% bromophenol blue
- The SDS compound present in the Laemmli SDS sample buffer binds noncovalently to proteins. SDS is negatively charged and can mask the intrinsic charge of the protein it binds
Application
- Laemmli SDS sample buffer is used to denature proteins and for protein sample preparation to be used in the SDS-PAGE system
- This sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of the running gel
- It is also used for polyacrylamide protein gel analysis

Specifications
Specifications
Color | Blue |
Concentration | 6X |
Product Type | Sample Loading Buffer |
Form | Liquid |
For Use With (Application) | Laemmli SDS-PAGE |
Quantity | 50 mL |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction