Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laforin/EPM2A Antibody, Novus Biologicals™
SDP

Catalog No. NBP224474 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224474 0.1 mg
NBP224474SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224474 Supplier Novus Biologicals Supplier No. NBP224474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Laforin/EPM2A Polyclonal specifically detects Laforin/EPM2A in Human, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Laforin/EPM2A
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.25-0.50 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q6VVB1
Gene Alias EC 3.1.3.16, EC 3.1.3.48, epilepsy, progressive myoclonus type 2, Lafora disease (laforin), epilepsy, progressive myoclonus type 2A, Lafora disease (laforin), EPM2, Lafora PTPase, laforin, LAFPTPase, LD, LDE, MELF
Gene Symbols EPM2A
Host Species Rabbit
Immunogen A portion of amino acids 80-130 of human Laforin/EPM2A was used as the immunogen for the antibody.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 7957
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, cow, horse, mouse, chimpanzee, dog and rat, 93% homologous in opossum, rhesus monkey, chicken, zebrafinch and Xenopus and 87% homologous in zebrafish and platypus.
Target Species Human, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.