Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laforin/EPM2A Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $426.00

Specifications

Antigen Laforin/EPM2A
Concentration 0.5 mg/mL
Dilution Western Blot 0.25-0.50 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224474SS
SDP
View Documents
Novus Biologicals
NBP224474SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224474
SDP
View Documents
Novus Biologicals
NBP224474
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

Laforin/EPM2A Polyclonal specifically detects Laforin/EPM2A in Human, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Laforin/EPM2A
Western Blot 0.25-0.50 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Rabbit
Protein Phosphatase
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
EC 3.1.3.16, EC 3.1.3.48, epilepsy, progressive myoclonus type 2, Lafora disease (laforin), epilepsy, progressive myoclonus type 2A, Lafora disease (laforin), EPM2, Lafora PTPase, laforin, LAFPTPase, LD, LDE, MELF
EPM2A
IgG
Affinity Purified
The amino acid sequence used as immunogen for the antibody is 100% homologous in human, cow, horse, mouse, chimpanzee, dog and rat, 93% homologous in opossum, rhesus monkey, chicken, zebrafinch and Xenopus and 87% homologous in zebrafish and platypus.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Bovine
Q6VVB1
7957
A portion of amino acids 80-130 of human Laforin/EPM2A was used as the immunogen for the antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.