Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG-3 Antibody (11E3), FITC, Novus Biologicals™
SDP

Catalog No. NB001340 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001340 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001340 Supplier Novus Biologicals Supplier No. NBP197662F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAG-3 Monoclonal specifically detects LAG-3 in Human, Monkey samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen LAG-3
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 11000
Conjugate FITC
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD223, CD223 antigen, FDC, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC
Gene Symbols LAG3
Host Species Mouse
Immunogen Recombinant human LAG-3 (lymphocyte activation gene-3).
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3902
Test Specificity Recognizes the first N-terminal D1 domain of human and monkey LAG-3.
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.