Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG-3 Antibody (2561B), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB340035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340035 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340035 Supplier Novus Biologicals Supplier No. FAB10395MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LAG-3 Monoclonal antibody specifically detects LAG-3 in Cynomolgus Monkey samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen LAG-3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2561B
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey LAG-3, Val20-Leu450, Accession # XP_005570011.1
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3902
Target Species Cynomolgus Monkey
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.